missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. T091425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T091425G 25 g
T0914500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T091425G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T091425G
Only null left

Chemical Identifiers

CAS 429-42-5
Molecular Formula C16H36BF4N
Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
PubChem CID 67932
IUPAC Name tetrabutylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 161°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 329.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Tetrafluoroborate
EINECSNumber (2)-0186
TSCA Yes