accessibility menu, dialog, popup

UserName

CAS RN 429-42-5

IUPAC Name: tetrabutylazanium; tetrafluoroboranuide
Synonyms: tetrabutylammonium ion tetrafluoroborate
Molecular Weight (g/mol): 329.27
Molecular Formula: C16H36BF4N
InChi Key: NNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (5)