missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Perrhenate 98.0+%, TCI America™

Catalog No. p-7136967
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18031G 1 g
T18035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18031G Supplier TCI America Supplier No. T18031G

Chemical Identifiers

CAS 16385-59-4
Molecular Formula C16H36NO4Re
Molecular Weight (g/mol) 492.674
MDL Number MFCD00012037
InChI Key MTTXKKTWRLXAKW-UHFFFAOYSA-N
PubChem CID 11123851
IUPAC Name oxido(trioxo)rhenium;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-][Re](=O)(=O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 492.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Perrhenate