accessibility menu, dialog, popup

UserName

CAS RN 16385-59-4

IUPAC Name: tetrabutylazanium rheniumoylolate
Synonyms: tetrabutylammonium ion; tetraoxorhenate(1-)
Molecular Weight (g/mol): 492.67
Molecular Formula: C16H36NO4Re
InChi Key: MTTXKKTWRLXAKW-UHFFFAOYSA-N
SMILES: [O-][Re](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results