missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium nitrate, 98%

Catalog No. AC420131000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420131000 100 g Glass bottle
AC420130050 5 g Glass bottle
AC420130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC420131000 Supplier Thermo Scientific Chemicals Supplier No. 420131000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1941-27-1 | C16H36N2O3 | 304.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 1941-27-1
Molecular Formula C16H36N2O3
Molecular Weight (g/mol) 304.46
MDL Number MFCD00043202
InChI Key QHOKENWFMZXSEU-UHFFFAOYSA-N
Synonym tetrabutylammonium nitrate, tetra-n-butylammonium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate, tetrabutylazanium nitrate, ammonium, tetrabutyl-, nitrate, n-n-n-tributyl-1-butanaminium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate 1:1, tetrabutylammonium ion nitrate, tetrabutylammoniumnitrate, acmc-209exx
PubChem CID 16027
IUPAC Name tetrabutylazanium;nitrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[N+](=O)([O-])[O-]

Specifications

Melting Point 116°C to 120°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3(CH2)3)4NNO3
Quantity 100 g
Solubility Information Solubility in water: <2%
Formula Weight 304.46
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrabutylammonium nitrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-726-0
RTECSNumber BS5475000
TSCA TSCA

RUO – Research Use Only