accessibility menu, dialog, popup

UserName

CAS RN 1941-27-1

IUPAC Name: tetrabutylazanium nitrate
Synonyms: tetrabutylammonium ion nitrate
Molecular Weight (g/mol): 304.48
Molecular Formula: C16H36N2O3
InChi Key: QHOKENWFMZXSEU-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)