missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide Solution, MP Biomedical

Catalog No. ICN15210450
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15210450 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15210450 Supplier MP Biomedicals Inc Supplier No. 0215210450

Tetrabutylammonium Hydroxide Solution has a concentration of 40% in H2O and is a phase transfer catalyst.

  • Used as a curing accelerator for epoxy resins
  • Also used in preparation of dichlorocarbene from chloroform

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1, tetrabutylammonium hydroxide solution in water
PubChem CID 2723671
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Quantity 50 mL
Concentration or Composition (by Analyte or Components) 40% in H2O
Packaging Glass
Density 0.99 g/mL at 25 °C (Lit.)