missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide, MP Biomedicals™
$367.61 - $367.61
Chemical Identifiers
| CAS | 2052-49-5 |
|---|---|
| Molecular Formula | C16H37NO |
| Molecular Weight (g/mol) | 259.48 |
| MDL Number | MFCD00009425 |
| InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
| Synonym | tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 |
| PubChem CID | 2723671 |
| IUPAC Name | tetrabutylazanium hydroxide |
| SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15210483
|
MP Biomedicals Inc
0215210483 |
250 mL |
Each for $367.61
|
|
|||||
Chemical Identifiers
| 2052-49-5 | |
| 259.48 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| 2723671 | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
| C16H37NO | |
| MFCD00009425 | |
| tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
| tetrabutylazanium hydroxide |
Specifications
| 2052-49-5 | |
| 0.98g/mL | |
| MFCD00009425 | |
| tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 259.48 | |
| 259.474 | |
| Tetrabutylammonium Hydroxide |
| Colorless | |
| C16H37NO | |
| 250 mL | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| tetrabutylazanium hydroxide | |
| 2723671 | |
| Liquid |
Safety and Handling
EINECSNumber : 218-147-6
Recommended Storage : Store at room temperature