missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide (40% in Water), TCI America™

Catalog No. T168525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T168525G Supplier TCI America Supplier No. T168525G

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Yellow
Quantity 25 g
UN Number 3267
Formula Weight 259.48
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide (40% in Water)
EINECSNumber (2)-0186
RTECSNumber BS5425000
TSCA Yes