Learn More
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6 | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Supplier: Thermo Scientific Chemicals 123535000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
| Tetrabutylammonium hydroxide | |
| Colorless to Yellow | |
| 4°C | |
| C16H37NO | |
| [CH3(CH2)3]4NOH | |
| 500 mL | |
| 05,645; 11,500; 07,332 | |
| tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
| VDZOOKBUILJEDG-UHFFFAOYSA-M | |
| tetrabutylazanium;hydroxide | |
| 2723671 | |
| Solution |
| 2052-49-5,108-88-3,67-56-1 | |
| 0.8500g/mL | |
| Glass bottle | |
| 1.3775 | |
| MFCD00009425 | |
| 04,II,634 | |
| 0.85 | |
| Solubility in water: insoluble | |
| [OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 259.48 | |
| 259.47 |
RUO – Research Use Only