missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Chemical Name or Material | Tetrabutylammonium hydroxide |
| CAS | 2052-49-5, 108-88-3, 67-56-1 |
| Color | Colorless to Yellow |
| Density | 0.8500g/mL |
| Flash Point | 4°C |
| Packaging | Glass bottle |
| Molecular Formula | C16H37NO |
| Refractive Index | 1.3775 |
| Linear Formula | [CH3(CH2)3]4NOH |
| MDL Number | MFCD00009425 |
| Show More |
RUO – Research Use Only