missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogen Sulfate 98.0+%, TCI America™

Catalog No. p-7136452
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T083525G 25 g
T0835100G 100 g
T0835500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T083525G Supplier TCI America Supplier No. T083525G
Only null left

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name tetrabutylazanium hydrogen sulfate
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 173°C
Color White
Quantity 25 g
Formula Weight 339.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Hydrogen Sulfate
EINECSNumber (2)-0186
TSCA Yes