missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydrogen Sulfate, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor T129425GM
Specifications
| 32503-27-8 | |
| 170° to 175°C | |
| C16H37NO4S | |
| SHFJWMWCIHQNCP-UHFFFAOYSA-M | |
| tetrabutylazanium hydrogen sulfate | |
| 0.98 |
| 1 | |
| Amber Glass Bottle | |
| 25 g | |
| OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC | |
| 339.54 | |
| Reagent |
Chemical Identifiers
| 32503-27-8 | |
| 339.54 | |
| tetrabutylazanium hydrogen sulfate |
| C16H37NO4S | |
| SHFJWMWCIHQNCP-UHFFFAOYSA-M | |
| OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.