missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Tetrabutylammonium hexafluorophosphate (TBAHFP) is a quaternary ammonium salt, mainly used as electrolyte. It is soluble in solvents like liquid sulphur dioxide and methylene chloride.

$283.80 - $1025.85

Chemical Identifiers

MDL Number MFCD00011748
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013891
View Documents
Merck Emd Millipore
8687925G
25 g
Each for $283.80
Only null left
 
111013892
View Documents
Merck Emd Millipore
86879100G
100 g
Each for $1,025.85
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011748

Specifications

244°C to 246°C
25 g
MFCD00011748
387.43
Electrochemical
3109-63-5
(CH3 CH2 CH2 CH2)4 N(PF6)
Acetonitrile: 0.1 g/mL, clear, colorless
≥99.0% (CHN); ≥99.0%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'