missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hexafluorophosphate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013892
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013892 100 g
111013891 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013892 Supplier Merck Emd Millipore Supplier No. 86879100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetrabutylammonium hexafluorophosphate (TBAHFP) is a quaternary ammonium salt, mainly used as electrolyte. It is soluble in solvents like liquid sulphur dioxide and methylene chloride.

Chemical Identifiers

MDL Number MFCD00011748

Specifications

Melting Point 244°C to 246°C
CAS 3109-63-5
Physical Form Solid
Quantity 100 g
Linear Formula (CH3 CH2 CH2 CH2)4 N(PF6)
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless
Molecular Weight (g/mol) 387.43
Formula Weight 387.43
Percent Purity ≥99.0% (CHN); ≥99.0%
Grade Electrochemical
Chemical Name or Material Tetrabutylammonium Hexafluorophosphate
Show More Show Less