missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran), TCI America™

Catalog No. T133825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T133825ML 25 mL
T1338100ML 100 mL
T1338500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T133825ML Supplier TCI America Supplier No. T133825ML

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
IUPAC Name tetrabutylazanium fluoride
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Undesignated
UN Number 2924
Formula Weight 261.47
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Chemical Name or Material Tetrabutylammonium Fluoride (ca. 1mol/L in Tetrahydrofuran)
EINECSNumber (2)-0186
TSCA Yes
Recommended Storage Refrigerator