missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water

Catalog No. p-195732
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC201950050 5 mL
AC201951000 100 mL
AC201955000 500 mL
AC201950025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC201950050 Supplier Thermo Scientific Chemicals Supplier No. 201950050

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Specifications

Chemical Name or Material Tetrabutylammonium fluoride
Name Note 1M Solution in THF, Containing ca 5% Water
CAS 429-41-4, 109-99-9, 7732-18-5
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
MDL Number MFCD00011747
Fieser 04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,363
Density 0.8870g/mL
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;fluoride
Molecular Weight (g/mol) 261.47
PubChem CID 2724141
ChEBI CHEBI:51990
Formula Weight 261.46
Packaging Glass bottle
Specific Gravity 0.887
Quantity 5 mL
Flash Point −17°C
Show More Show Less

RUO – Research Use Only