missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Dibromoiodide 97.0+%, TCI America™

Supplier:  TCI America T12691G

 View more versions of this product

Catalog No. T12691G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium Dibromoiodide
81°C
MFCD00059121
LFZGDXLUTJIZAG-UHFFFAOYSA-N
529.183
529.18
Crystalline Powder
15802-00-3
C16H36Br2IN
1 g
CCCC[N+](CCCC)(CCCC)CCCC.Br[I-]Br
13867581
≥97.0% (T)

Chemical Identifiers

15802-00-3
529.183
LFZGDXLUTJIZAG-UHFFFAOYSA-N
CCCC[N+](CCCC)(CCCC)CCCC.Br[I-]Br
C16H36Br2IN
MFCD00059121
13867581
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : No

SDS
Documents

Documents

Promotions

Promotions