missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium chloride, 95%, Tech., contains also bromide

Catalog No. p-3737447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174120050 5 g Glass bottle
AC174120250 25 g Glass bottle
AC174121000 100 g Glass bottle
AC174125000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC174120050 Supplier Thermo Scientific Chemicals Supplier No. 174120050
Only null left

CAS: 1112-67-0 | C16H36ClN | 277.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 1112-67-0
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name tetrabutylazanium;chloride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Cl-]

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NCl
Quantity 5 g
Beilstein 04, III, 292
Fieser 06,565; 07,353; 17,363
Solubility Information Solubility in water: soluble.
Formula Weight 277.92
Percent Purity 95%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material Tetrabutylammonium chloride, Also contains bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 214-195-7

RUO – Research Use Only