missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Borohydride 96.0+%, TCI America™

Supplier:  TCI America T091725G

 View more versions of this product

Catalog No. T091725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium Borohydride [Reducing Reagent]
126°C
C16H40BN
25 g
tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
[BH4-].CCCC[N+](CCCC)(CCCC)CCCC
257.31
257.31
Crystalline Powder
33725-74-5
White
MFCD00012035
3131
GMBOFJFPOCGSOI-UHFFFAOYSA-N
tetrabutylazanium boranuide
9881569
≥96.0% (T)

Chemical Identifiers

33725-74-5
257.31
GMBOFJFPOCGSOI-UHFFFAOYSA-N
9881569
[BH4-].CCCC[N+](CCCC)(CCCC)CCCC
C16H40BN
MFCD00012035
tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
tetrabutylazanium boranuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186&(2)-3044

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions