missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Borohydride 96.0+%, TCI America™
$61.51 - $175.22
Chemical Identifiers
| CAS | 33725-74-5 |
|---|---|
| Molecular Formula | C16H40BN |
| Molecular Weight (g/mol) | 257.31 |
| MDL Number | MFCD00012035 |
| InChI Key | GMBOFJFPOCGSOI-UHFFFAOYSA-N |
| Synonym | tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium |
| PubChem CID | 9881569 |
| IUPAC Name | tetrabutylazanium boranuide |
| SMILES | [BH4-].CCCC[N+](CCCC)(CCCC)CCCC |
Chemical Identifiers
| 33725-74-5 | |
| 257.31 | |
| GMBOFJFPOCGSOI-UHFFFAOYSA-N | |
| 9881569 | |
| [BH4-].CCCC[N+](CCCC)(CCCC)CCCC |
| C16H40BN | |
| MFCD00012035 | |
| tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium | |
| tetrabutylazanium boranuide |
Specifications
| 33725-74-5 | |
| White | |
| MFCD00012035 | |
| 3131 | |
| GMBOFJFPOCGSOI-UHFFFAOYSA-N | |
| tetrabutylazanium boranuide | |
| 9881569 | |
| ≥96.0% (T) | |
| Tetrabutylammonium Borohydride [Reducing Reagent] |
| 126°C | |
| C16H40BN | |
| 5 g | |
| tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium | |
| [BH4-].CCCC[N+](CCCC)(CCCC)CCCC | |
| 257.31 | |
| 257.31 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186&(2)-3044
TSCA : Yes