missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutyl Orthotitanate Tetramer, TCI America™

Catalog No. B074325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B074325G Supplier TCI America Supplier No. B074325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70799-68-7
Molecular Formula C40H90O13Ti4
Molecular Weight (g/mol) 970.615
MDL Number MFCD00070567
InChI Key LEAWWEHOHYFEPH-UHFFFAOYSA-N
Synonym Butyl Titanate Tetramer, Tetrabutoxy Titan Tetramer, Tetrabutyl Titanate Tetramer, Titanium Butoxide Tetramer
PubChem CID 53384475
IUPAC Name butan-1-olate;oxygen(2-);titanium(4+)
SMILES CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-].[O-2].[O-2].[O-2].[Ti+4].[Ti+4].[Ti+4].[Ti+4]

Specifications

Color Yellow
Quantity 25 g
UN Number 1993
Formula Weight 970.62
Physical Form Liquid
Chemical Name or Material Tetrabutyl Orthotitanate Tetramer