accessibility menu, dialog, popup

UserName

CAS RN 70799-68-7

H3COH3COH3COH3COCH3OCH3OCH3OCH3OCH3OCH3OOOOTiTiTiTi

CAS RN 70799-68-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results