missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromothiophene 99.0+%, TCI America™

Catalog No. p-7136633
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11495G 5 g
T114925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11495G Supplier TCI America Supplier No. T11495G
Only null left

Chemical Identifiers

CAS 3958-03-0
Molecular Formula C4Br4S
Molecular Weight (g/mol) 399.72
MDL Number MFCD00005419
InChI Key AVPWUAFYDNQGNZ-UHFFFAOYSA-N
Synonym tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen
PubChem CID 77565
IUPAC Name 2,3,4,5-tetrabromothiophene
SMILES C1(=C(SC(=C1Br)Br)Br)Br

Specifications

Melting Point 115°C
Boiling Point 173°C
Quantity 5 g
Formula Weight 399.72
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromothiophene