Learn More
Tetrabromothiophene, 99%
CAS: 3958-03-0 | C4Br4S | 399.72 g/mol
Supplier: Thermo Scientific Chemicals A1300922
Description
Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases, strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications
| Tetrabromothiophene | |
| 113°C to 117°C | |
| Odorless | |
| MFCD00005419 | |
| 383664 | |
| Insoluble in water. Soluble in organic solvents. | |
| C1(=C(SC(=C1Br)Br)Br)Br | |
| 399.72 | |
| 399.72 |
| 3958-03-0 | |
| 324°C to 327°C | |
| C4Br4S | |
| 100 g | |
| tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen | |
| AVPWUAFYDNQGNZ-UHFFFAOYSA-N | |
| 2,3,4,5-tetrabromothiophene | |
| 77565 | |
| 99% |
Chemical Identifiers
| 3958-03-0 | |
| 399.72 | |
| AVPWUAFYDNQGNZ-UHFFFAOYSA-N | |
| 77565 | |
| C1(=C(SC(=C1Br)Br)Br)Br |
| C4Br4S | |
| MFCD00005419 | |
| tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen | |
| 2,3,4,5-tetrabromothiophene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-554-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only