missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromothiophene, 99%

Catalog No. AAA1300914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1300914 25 g
AAA1300922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1300914 Supplier Thermo Scientific Chemicals Supplier No. A1300914

CAS: 3958-03-0 | C4Br4S | 399.72 g/mol

Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromothiophene, holds wide applications in organic electronics and photonics. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases, strong oxidizing agents. Stable under normal temperatures and pressures.

Chemical Identifiers

CAS 3958-03-0
Molecular Formula C4Br4S
Molecular Weight (g/mol) 399.72
MDL Number MFCD00005419
InChI Key AVPWUAFYDNQGNZ-UHFFFAOYSA-N
Synonym tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen
PubChem CID 77565
IUPAC Name 2,3,4,5-tetrabromothiophene
SMILES C1(=C(SC(=C1Br)Br)Br)Br

Specifications

Melting Point 113°C to 117°C
Boiling Point 324°C to 327°C
Odor Odorless
Quantity 25 g
Beilstein 383664
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 399.72
Percent Purity 99%
Chemical Name or Material Tetrabromothiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-554-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only