missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophthalic Anhydride 95.0+%, TCI America™

Catalog No. p-7136050
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
T004625G 25 g
T0046100G 100 g
2 options

Catalog No. T004625G

Supplier: TCI America T004625G

Only null left

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8Br4O3
Molecular Weight (g/mol) 463.701
MDL Number MFCD00005919
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
PubChem CID 12443
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 25 g
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Formula Weight 463.70
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromophthalic Anhydride
EINECSNumber (3)-0062
RTECSNumber TI3350000
TSCA Yes