missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenolphthalein ethyl ester, pure

CAS: 1176-74-5 | C22H14Br4O4 | 661.96 g/mol

$709.37 - $709.37

Chemical Identifiers

CAS 1176-74-5
Molecular Formula C22H14Br4O4
Molecular Weight (g/mol) 661.96
MDL Number MFCD00066387
InChI Key SQFXATUXPUCFFO-UHFFFAOYSA-N
Synonym tetrabromophenolphthalein ethyl ester, 3',3,5',5-tetrabromophenolphthalein ethyl ester, ethyl tetrabromophenolphthalein, unii-6d91ns781o, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl benzoate, 3 inverted exclamation marka,3 inverted exclamation marka,5 inverted exclamation marka,5 inverted exclamation marka-tetrabromophenolphthalein ethyl ester, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, benzoic acid,2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-,ethyl ester, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene methyl benzoate
PubChem CID 92873
IUPAC Name ethyl 2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
SMILES CCOC(=O)C1=CC=CC=C1C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC420070010
SDS View Documents
Thermo Scientific Chemicals
420070010
1 g Glass bottle
Each for $709.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1176-74-5
661.96
SQFXATUXPUCFFO-UHFFFAOYSA-N
92873
CCOC(=O)C1=CC=CC=C1C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br
C22H14Br4O4
MFCD00066387
tetrabromophenolphthalein ethyl ester, 3',3,5',5-tetrabromophenolphthalein ethyl ester, ethyl tetrabromophenolphthalein, unii-6d91ns781o, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl benzoate, 3 inverted exclamation marka,3 inverted exclamation marka,5 inverted exclamation marka,5 inverted exclamation marka-tetrabromophenolphthalein ethyl ester, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, benzoic acid,2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-,ethyl ester, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene methyl benzoate
ethyl 2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate

Specifications

1176-74-5
Red to Yellow
Glass bottle
MFCD00066387
15, 9331
Solubility in water: negligible.
SQFXATUXPUCFFO-UHFFFAOYSA-N
ethyl 2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
92873
Pure
Tetrabromophenolphthalein ethyl ester
208.0°C to 211.0°C
Authentic
C22H14Br4O4
1 g
tetrabromophenolphthalein ethyl ester, 3',3,5',5-tetrabromophenolphthalein ethyl ester, ethyl tetrabromophenolphthalein, unii-6d91ns781o, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl benzoate, 3 inverted exclamation marka,3 inverted exclamation marka,5 inverted exclamation marka,5 inverted exclamation marka-tetrabromophenolphthalein ethyl ester, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, benzoic acid,2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-,ethyl ester, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene methyl benzoate
80.0% min.
CCOC(=O)C1=CC=CC=C1C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br
661.96
661.96
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-645-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only