missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromocatechol, 96%

Catalog No. AAL0069006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0069006 5 g
AAL0069014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0069006 Supplier Thermo Scientific Chemicals Supplier No. L0069006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 488-47-1 | C6H2Br4O2 | 425.70 g/mol

Tetrabromocatechol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromocatechol is used as a pharmaceutical intermediate.

Solubility
It is partly miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.

Chemical Identifiers

CAS 488-47-1
Molecular Formula C6H2Br4O2
Molecular Weight (g/mol) 425.70
MDL Number MFCD00002189
InChI Key OAUWOBSDSJNJQP-UHFFFAOYSA-N
Synonym tetrabromocatechol, tetrabromopyrocatechol, pyrocatechol, tetrabromo, 1,2-benzenediol, 3,4,5,6-tetrabromo, 3,4,5,6-tetrabromocatechol, unii-n642cph23m, 1, 3,4,5,6-tetrabromo, 4-06-00-05624 beilstein handbook reference
PubChem CID 61127
IUPAC Name 3,4,5,6-tetrabromobenzene-1,2-diol
SMILES OC1=C(O)C(Br)=C(Br)C(Br)=C1Br

Specifications

Melting Point 189°C to 193°C
Quantity 5 g
Beilstein 2051548
Solubility Information It is partly miscible with water.
Formula Weight 425.7
Percent Purity 96%
Chemical Name or Material Tetrabromocatechol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-678-9
RTECSNumber UX2430000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only