missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromo-o-sulfobenzoic Anhydride 98.0+%, TCI America™

Catalog No. T00485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00485G 5 g
T004825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00485G Supplier TCI America Supplier No. T00485G
Only null left

Chemical Identifiers

CAS 68460-01-5
Molecular Formula C7Br4O4S
Molecular Weight (g/mol) 499.749
MDL Number MFCD00009745
InChI Key QPGYGIVRJIICGZ-UHFFFAOYSA-N
PubChem CID 110335
IUPAC Name 4,5,6,7-tetrabromo-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)S(=O)(=O)OC2=O

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 499.75
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromo-o-sulfobenzoic Anhydride