accessibility menu, dialog, popup

UserName

BrBrBrBrOOSOO

CAS RN 68460-01-5

IUPAC Name: tetrabromo-3H-2,1λ⁶-benzoxathiole-1,1,3-trione
Synonyms: tetrabromo-2,1λ⁶-benzoxathiole-1,1,3-trione
Molecular Weight (g/mol): 499.75
Molecular Formula: C7Br4O4S
InChi Key: QPGYGIVRJIICGZ-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OS(=O)(=O)C2=C(Br)C(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results