missing translation for 'onlineSavingsMsg'
Learn More

Tetraamylammonium Iodide 98.0+%, TCI America™

Catalog No. p-7136550
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10115G 5 g
T101125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10115G Supplier TCI America Supplier No. T10115G
Only null left

Chemical Identifiers

CAS 2498-20-6
Molecular Formula C20H44IN
Molecular Weight (g/mol) 425.483
MDL Number MFCD00041980
InChI Key FBLZDUAOBOMSNZ-UHFFFAOYSA-M
Synonym tetrapentylammonium iodide, tetraamylammonium iodide, tetra-n-pentylammonium iodide, 1-pentanaminium, n,n,n-tripentyl-, iodide 1:1, tetrapentylazanium iodide, tetra-n-amylammonium iodide, tetraamylammoniumjodid, tetraamylammoniumiodide, acmc-209ggb, 1-pentanaminium, n,n,n-tripentyl-, iodide
PubChem CID 17248
IUPAC Name tetrapentylazanium;iodide
SMILES CCCCC[N+](CCCCC)(CCCCC)CCCCC.[I-]

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 425.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraamylammonium Iodide
EINECSNumber (2)-0186
TSCA Yes