accessibility menu, dialog, popup

UserName

CAS RN 2498-20-6

IUPAC Name: tetrapentylazanium iodide
Synonyms: tetrapentylammonium iodide
Molecular Weight (g/mol): 425.48
Molecular Formula: C20H44IN
InChi Key: FBLZDUAOBOMSNZ-UHFFFAOYSA-M
SMILES: [I-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)