missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraamylammonium Chloride 98.0+%, TCI America™
$275.87 - $814.59
Chemical Identifiers
| CAS | 4965-17-7 |
|---|---|
| Molecular Formula | C20H44ClN |
| Molecular Weight (g/mol) | 334.029 |
| MDL Number | MFCD00011857 |
| InChI Key | SXAWRMKQZKPHNJ-UHFFFAOYSA-M |
| Synonym | Tetrapentylammonium Chloride |
| PubChem CID | 78667 |
| IUPAC Name | tetrapentylazanium;chloride |
| SMILES | CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-] |
Chemical Identifiers
| 4965-17-7 | |
| 334.029 | |
| SXAWRMKQZKPHNJ-UHFFFAOYSA-M | |
| 78667 | |
| CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-] |
| C20H44ClN | |
| MFCD00011857 | |
| Tetrapentylammonium Chloride | |
| tetrapentylazanium;chloride |
Specifications
| 4965-17-7 | |
| C20H44ClN | |
| 5 g | |
| Tetrapentylammonium Chloride | |
| CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-] | |
| 334.029 | |
| 334.03 | |
| Crystalline Powder |
| White | |
| MFCD00011857 | |
| 2811 | |
| SXAWRMKQZKPHNJ-UHFFFAOYSA-M | |
| tetrapentylazanium;chloride | |
| 78667 | |
| ≥98.0% (T) | |
| Tetraamylammonium Chloride |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : RZ9275000
TSCA : Yes