accessibility menu, dialog, popup

UserName

ClH3CNCH3CH3CH3

CAS RN 4965-17-7

IUPAC Name: tetrapentylazanium chloride
Synonyms: tetrapentylammonium chloride
Molecular Weight (g/mol): 334.03
Molecular Formula: C20H44ClN
InChi Key: SXAWRMKQZKPHNJ-UHFFFAOYSA-M
SMILES: [Cl-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results