missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraamylammonium Bromide 98.0+%, TCI America™
Supplier: TCI America T143225G
Specifications
| Tetraamylammonium Bromide | |
| 101°C | |
| C20H44BrN | |
| 25 g | |
| SPALIFXDWQTXKS-UHFFFAOYSA-M | |
| tetrapentylazanium bromide | |
| 70086 | |
| ≥98.0% (T) |
| 866-97-7 | |
| White | |
| MFCD00011856 | |
| tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide | |
| [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC | |
| 378.48 | |
| 378.48 | |
| Crystalline Powder |
Chemical Identifiers
| 866-97-7 | |
| 378.48 | |
| SPALIFXDWQTXKS-UHFFFAOYSA-M | |
| 70086 | |
| [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC |
| C20H44BrN | |
| MFCD00011856 | |
| tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide | |
| tetrapentylazanium bromide |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes