missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraammineplatinum(II) tetrachloroplatinate(II), Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | 320°C (decomposition) |
| Density | 4.000 g/mL |
| Quantity | 1 g |
| Formula Weight | 600.12 |
| Physical Form | Powder |
| Chemical Name or Material | Tetraammineplatinum(II) tetrachloroplatinate(II) |
Safety and Handling
| Hazard Category | H301-H317-H318-H334 |
| Hazard Statement | GHS H Statement H301-H334-H318-H317 Toxic if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P272-P280-P285-P301+P310-P302+P352-P304+P341-P305+P351+P338-P310-P330-P333+P313-P342+P311-P363-P501c |
| EINECSNumber | 237-501-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |