accessibility menu, dialog, popup

UserName

CAS RN 13820-46-7

PtNNClPtClClClNN

CAS RN 13820-46-7

IUPAC Name: platinum(2+) tetrachloroplatinum tetraamine
Synonyms: platinum(2+) platinum tetrachloride tetraamine
Molecular Weight (g/mol): 600.09
Molecular Formula: Cl4H12N4Pt2
InChi Key: LYCTXVQKJYMIAX-UHFFFAOYSA-J
SMILES: N.N.N.N.[Pt++].Cl[Pt](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results