missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydrogen sulfate, 98%

Catalog No. p-7081838
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1200206 5 g
AAA1200214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1200206 Supplier Thermo Scientific Chemicals Supplier No. A1200206

CAS: 56211-70-2 | C12H29NO4S | 283.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56211-70-2
Molecular Formula C12H29NO4S
Molecular Weight (g/mol) 283.43
MDL Number MFCD00038764
InChI Key MOXJKKOSZCHGEU-UHFFFAOYSA-M
Synonym tetrapropylammonium bisulfate, tetra-n-propylammonium hydrogen sulfate, tetrapropylammonium hydrogen sulfate, hydrogen sulfate; tetrapropylammonium, tetrapropyl ammonium hydrogen sulphate, acmc-1avly, hydrogen sulfate; tetrapropylazanium, n,n,n-tripropylpropan-1-aminium hydrogen sulfate, 1-propanaminium, n,n,n-tripropyl-, sulfate
PubChem CID 16218669
IUPAC Name hydrogen sulfate;tetrapropylazanium
SMILES OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC

Specifications

Melting Point 158°C to 160°C
Linear Formula (CH3CH2CH2)4NHSO4
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 283.43
Percent Purity 98%
Chemical Name or Material Tetra-n-propylammonium hydrogen sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only