CAS RN 56211-70-2
IUPAC Name:
tetrapropylazanium hydrogen sulfate
Synonyms:
tetrapropylammonium hydrogen sulfate
Molecular Weight (g/mol):
283.43
Molecular Formula:
C12H29NO4S
InChi Key:
MOXJKKOSZCHGEU-UHFFFAOYSA-M
SMILES:
OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC
1
–
1
of
1
results
1
Promotions Available
Tetra-n-propylammonium hydrogen sulfate, 98%
Tetra-n-propylammonium hydrogen sulfate, 98%
CAS: 56211-70-2 Molecular Formula: C12H29NO4S Molecular Weight (g/mol): 283.43 MDL Number: MFCD00038764 InChI Key: MOXJKKOSZCHGEU-UHFFFAOYSA-M Synonym: tetrapropylammonium bisulfate,tetra-n-propylammonium hydrogen sulfate,tetrapropylammonium hydrogen sulfate,hydrogen sulfate; tetrapropylammonium,tetrapropyl ammonium hydrogen sulphate,acmc-1avly,hydrogen sulfate; tetrapropylazanium,n,n,n-tripropylpropan-1-aminium hydrogen sulfate,1-propanaminium, n,n,n-tripropyl-, sulfate PubChem CID: 16218669 IUPAC Name: hydrogen sulfate;tetrapropylazanium SMILES: OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC