Learn More
Tetra-n-propoxysilane, 97%
CAS: 682-01-9 | C12H28O4Si | 264.437 g/mol
$70.26 - $165.05
Chemical Identifiers
| CAS | 682-01-9 |
|---|---|
| Molecular Formula | C12H28O4Si |
| Molecular Weight (g/mol) | 264.437 |
| MDL Number | MFCD00009364 |
| InChI Key | ZQZCOBSUOFHDEE-UHFFFAOYSA-N |
| Synonym | tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane |
| PubChem CID | 69627 |
| IUPAC Name | tetrapropyl silicate |
| SMILES | CCCO[Si](OCCC)(OCCC)OCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1664414
|
Thermo Scientific Chemicals
L1664414 |
25 g |
Each for $70.26
|
|
|||||
|
AAL1664422
|
Thermo Scientific Chemicals
L1664422 |
100 g |
Each for $165.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 682-01-9 | |
| 264.437 | |
| ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
| 69627 | |
| CCCO[Si](OCCC)(OCCC)OCCC |
| C12H28O4Si | |
| MFCD00009364 | |
| tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
| tetrapropyl silicate |
Specifications
| 224°C to 225°C (94°C (5mmHg)) | |
| 25 g | |
| (CH3CH2CH2O)4Si | |
| 1706115 | |
| tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
| CCCO[Si](OCCC)(OCCC)OCCC | |
| 264.437 | |
| 264.44 | |
| 1.401 | |
| 0.916 |
| 682-01-9 | |
| C12H28O4Si | |
| MFCD00009364 | |
| 95°C (203°F) | |
| ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
| tetrapropyl silicate | |
| 69627 | |
| 97% | |
| Moisture sensitive | |
| Tetra-n-propoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-659-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only