missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propoxysilane, 97%

CAS: 682-01-9 | C12H28O4Si | 264.437 g/mol

Supplier:  Thermo Scientific Chemicals L1664422

 View more versions of this product

Catalog No. AAL1664422


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

224°C to 225°C (94°C (5mmHg))
682-01-9
C12H28O4Si
MFCD00009364
95°C (203°F)
ZQZCOBSUOFHDEE-UHFFFAOYSA-N
tetrapropyl silicate
69627
97%
Moisture sensitive
Tetra-n-propoxysilane
100 g
(CH3CH2CH2O)4Si
1706115
tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane
CCCO[Si](OCCC)(OCCC)OCCC
264.437
264.44
1.401
0.916

Chemical Identifiers

682-01-9
264.437
ZQZCOBSUOFHDEE-UHFFFAOYSA-N
69627
CCCO[Si](OCCC)(OCCC)OCCC
C12H28O4Si
MFCD00009364
tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane
tetrapropyl silicate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-659-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only