missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-pentylammonium iodide, 98%

Catalog No. p-7046366
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1408814 25 g
AAL1408822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1408814 Supplier Thermo Scientific Chemicals Supplier No. L1408814
Only null left

CAS: 2498-20-6 | C20H44IN | 425.483 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2498-20-6
Molecular Formula C20H44IN
Molecular Weight (g/mol) 425.483
MDL Number MFCD00041980
InChI Key FBLZDUAOBOMSNZ-UHFFFAOYSA-M
Synonym tetrapentylammonium iodide, tetraamylammonium iodide, tetra-n-pentylammonium iodide, 1-pentanaminium, n,n,n-tripentyl-, iodide 1:1, tetrapentylazanium iodide, tetra-n-amylammonium iodide, tetraamylammoniumjodid, tetraamylammoniumiodide, acmc-209ggb, 1-pentanaminium, n,n,n-tripentyl-, iodide
PubChem CID 17248
IUPAC Name tetrapentylazanium;iodide
SMILES CCCCC[N+](CCCCC)(CCCCC)CCCCC.[I-]

Specifications

Melting Point 135°C to 139°C
Linear Formula [CH3(CH2)4]4NI
Quantity 25 g
Beilstein 3917412
Sensitivity Light Sensitive
Formula Weight 425.49
Percent Purity 98%
Chemical Name or Material Tetra-n-pentylammonium iodide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 219-688-0
TSCA Yes

RUO – Research Use Only