missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-octylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7136868
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T160310G 10 g
T160325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T160310G Supplier TCI America Supplier No. T160310G
Only null left

Chemical Identifiers

CAS 14866-33-2
Molecular Formula C32H68BrN
Molecular Weight (g/mol) 546.807
MDL Number MFCD00011863
InChI Key QBVXKDJEZKEASM-UHFFFAOYSA-M
Synonym tetraoctylammonium bromide, tetra-n-octylammonium bromide, tetraoctylazanium bromide, tetraoctyl ammonium bromide, 1-octanaminium, n,n,n-trioctyl-, bromide, unii-52qw29owoc, 52qw29owoc, tetra n-octyl ammonium bromide, acmc-1bvth, tetra-octylammonium bromide
PubChem CID 2734117
IUPAC Name tetraoctylazanium;bromide
SMILES CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]

Specifications

Melting Point 97°C
Color White
Quantity 10 g
Formula Weight 546.81
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetra-n-octylammonium Bromide
EINECSNumber (9)-1971
TSCA No