accessibility menu, dialog, popup

UserName

BrCH3NCH3CH3CH3

CAS RN 14866-33-2

IUPAC Name: tetraoctylazanium bromide
Synonyms: tetraoctylammonium bromide
Molecular Weight (g/mol): 546.81
Molecular Formula: C32H68BrN
InChi Key: QBVXKDJEZKEASM-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)