missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-heptylammonium bromide, 99%

CAS: 4368-51-8 | C28H60BrN | 490.699 g/mol

Supplier:  Thermo Scientific Chemicals A1521814

 View more versions of this product

Catalog No. AAA1521814


Only null left
Explore available promotions

Description

Description

Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetra-n-heptylammonium bromide
87°C to 89°C
MFCD00011861
Hygroscopic
Soluble in water and methanol.
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]
490.699
490.7
4368-51-8
C28H60BrN
25 g
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
YQIVQBMEBZGFBY-UHFFFAOYSA-M
tetraheptylazanium;bromide
78073
99%

Chemical Identifiers

4368-51-8
490.699
YQIVQBMEBZGFBY-UHFFFAOYSA-M
78073
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]
C28H60BrN
MFCD00011861
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
tetraheptylazanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-459-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only