missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-heptylammonium bromide, 99%

Catalog No. p-7022435
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1521806 5 g
AAA1521814 25 g
AAA1521822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1521806 Supplier Thermo Scientific Chemicals Supplier No. A1521806
Only null left

CAS: 4368-51-8 | C28H60BrN | 490.699 g/mol

Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4368-51-8
Molecular Formula C28H60BrN
Molecular Weight (g/mol) 490.699
MDL Number MFCD00011861
InChI Key YQIVQBMEBZGFBY-UHFFFAOYSA-M
Synonym tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
PubChem CID 78073
IUPAC Name tetraheptylazanium;bromide
SMILES CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]

Specifications

Melting Point 87°C to 89°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
Formula Weight 490.7
Percent Purity 99%
Chemical Name or Material Tetra-n-heptylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-459-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only