Learn More
Tetra-n-butylphosphonium iodide, 98%
CAS: 3115-66-0 | C16H36IP | 386.34 g/mol
Supplier: Thermo Scientific Chemicals A1679214
Description
Tetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.
Solubility
Soluble in water.
Notes
Hygroscopic. Store at 4°C. Store in the dark. Incompatible with oxidizing agents and light.
Specifications
| Tetra-n-butylphosphonium iodide | |
| 98°C to 100°C | |
| [CH3(CH2)3]4PI | |
| 25 g | |
| tetrabutylphosphonium iodide, tetra-n-butylphosphonium iodide, phosphonium, tetrabutyl-, iodide, tetrabutylphosphanium iodide, acmc-1crr0, wln: 4p4&4&4 &i, phosphonium,tetrabutyl-, iodide 1:1 | |
| CCIYPTIBRAUPLQ-UHFFFAOYSA-M | |
| tetrabutylphosphanium;iodide | |
| 201022 | |
| 98% |
| 3115-66-0 | |
| C16H36IP | |
| MFCD00050246 | |
| Hygroscopic | |
| Soluble in water. | |
| [I-].CCCC[P+](CCCC)(CCCC)CCCC | |
| 386.34 | |
| 386.34 |
Chemical Identifiers
| 3115-66-0 | |
| 386.34 | |
| CCIYPTIBRAUPLQ-UHFFFAOYSA-M | |
| 201022 | |
| [I-].CCCC[P+](CCCC)(CCCC)CCCC |
| C16H36IP | |
| MFCD00050246 | |
| tetrabutylphosphonium iodide, tetra-n-butylphosphonium iodide, phosphonium, tetrabutyl-, iodide, tetrabutylphosphanium iodide, acmc-1crr0, wln: 4p4&4&4 &i, phosphonium,tetrabutyl-, iodide 1:1 | |
| tetrabutylphosphanium;iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : TA2420000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only