CAS RN 3115-66-0
IUPAC Name:
tetrabutylphosphanium iodide
Synonyms:
tetrabutylphosphonium iodide
Molecular Weight (g/mol):
386.34
Molecular Formula:
C16H36IP
InChi Key:
CCIYPTIBRAUPLQ-UHFFFAOYSA-M
SMILES:
[I-].CCCC[P+](CCCC)(CCCC)CCCC
1
–
1
of
1
results
1
Promotions Available
Tetra-n-butylphosphonium iodide, 98%
Tetra-n-butylphosphonium iodide, 98%
CAS: 3115-66-0 Molecular Formula: C16H36IP Molecular Weight (g/mol): 386.34 MDL Number: MFCD00050246 InChI Key: CCIYPTIBRAUPLQ-UHFFFAOYSA-M Synonym: tetrabutylphosphonium iodide,tetra-n-butylphosphonium iodide,phosphonium, tetrabutyl-, iodide,tetrabutylphosphanium iodide,acmc-1crr0,wln: 4p4&4&4 &i,phosphonium,tetrabutyl-, iodide 1:1 PubChem CID: 201022 IUPAC Name: tetrabutylphosphanium;iodide SMILES: [I-].CCCC[P+](CCCC)(CCCC)CCCC