missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylphosphonium iodide, 98%

Catalog No. AAA1679206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1679206 5 g
AAA1679214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1679206 Supplier Thermo Scientific Chemicals Supplier No. A1679206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3115-66-0 | C16H36IP | 386.34 g/mol

Tetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.

Solubility
Soluble in water.

Notes
Hygroscopic. Store at 4°C. Store in the dark. Incompatible with oxidizing agents and light.

Chemical Identifiers

CAS 3115-66-0
Molecular Formula C16H36IP
Molecular Weight (g/mol) 386.34
MDL Number MFCD00050246
InChI Key CCIYPTIBRAUPLQ-UHFFFAOYSA-M
Synonym tetrabutylphosphonium iodide, tetra-n-butylphosphonium iodide, phosphonium, tetrabutyl-, iodide, tetrabutylphosphanium iodide, acmc-1crr0, wln: 4p4&4&4 &i, phosphonium,tetrabutyl-, iodide 1:1
PubChem CID 201022
IUPAC Name tetrabutylphosphanium;iodide
SMILES [I-].CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 98°C to 100°C
Linear Formula [CH3(CH2)3]4PI
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 386.34
Percent Purity 98%
Chemical Name or Material Tetra-n-butylphosphonium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber TA2420000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only